Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000594.3 | NP_000585.2 | 4 | 847 | 59 | |
GenBank mRNA | AB451492.1 | - | 2 | 444 | 59 | |
AF043342.1 | - | 2 | 444 | 59 | ||
AK314960.1 | - | 4 | 846 | 59 | ||
BC028148.1 | - | 4 | 829 | 5217636 | 59 | |
FJ795028.1 | - | 4 | 671 | 59 | ||
HQ201306.2 | - | 4 | 672 | 59 | ||
M10988.1 | - | 4 | 757 | 59 | ||
X01394.1 | - | 4 | 824 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
