Assay Details
Gene Symbol: FXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013438.2 | NP_001013456.1 | 3 | 390 | 83 | |
NM_001013439.2 | NP_001013457.1 | 4 | 734 | 83 | ||
NM_005087.3 | NP_005078.2 | 3 | 390 | 83 | ||
XM_005247813.3 | XP_005247870.2 | 3 | 165 | 83 | ||
XM_005247814.3 | XP_005247871.2 | 3 | 165 | 83 | ||
XM_005247815.3 | XP_005247872.2 | 3 | 165 | 83 | ||
XM_005247816.3 | XP_005247873.2 | 3 | 165 | 83 | ||
XM_006713775.3 | XP_006713838.1 | 4 | 508 | 83 | ||
XM_017007298.1 | XP_016862787.1 | 4 | 508 | 83 | ||
XM_017007299.1 | XP_016862788.1 | 4 | 508 | 83 | ||
XM_017007300.1 | XP_016862789.1 | 4 | 508 | 83 | ||
XM_017007301.1 | XP_016862790.1 | 4 | 508 | 83 | ||
GenBank mRNA | AK225334.1 | - | 3 | 166 | 83 | |
AK292633.1 | - | 3 | 166 | 83 | ||
AK294603.1 | - | 2 | 112 | 83 | ||
AK302197.1 | - | 3 | 169 | 83 | ||
AU280585.1 | - | 4 | 509 | 83 | ||
AY341428.1 | - | 3 | 441 | 83 | ||
BC028983.1 | - | 3 | 160 | 5122574 | 83 | |
BX956500.1 | - | 3 | 83 | |||
DC352767.1 | - | 3 | 390 | 83 | ||
KR259877.1 | - | 3 | 150 | 83 | ||
KR259878.1 | - | 3 | 150 | 83 | ||
KR259879.1 | - | 3 | 150 | 83 | ||
KR259880.1 | - | 4 | 494 | 83 | ||
U25165.1 | - | 3 | 162 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK294603.1, AY341428.1, NM_001013439.2, XM_006713775.3, XM_017007298.1, XM_017007299.1, XM_017007300.1, XM_017007301.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
