Assay Details
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286774.1 | NP_001273703.1 | 2 - 3 | 300 | 70 | |
GenBank mRNA | BU508058.1 | - | 2 - 3 | 188 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286774.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
