Assay Details
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286774.1 | NP_001273703.1 | 3 - 4 | 352 | 100 | |
GenBank mRNA | BU508058.1 | - | 3 - 4 | 240 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286774.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
