Assay Details
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000310.3 | NP_000301.1 | 3 - 4 | 596 | 104 | |
XM_005271008.1 | XP_005271065.1 | 3 - 4 | 380 | 104 | ||
GenBank mRNA | AK301682.1 | - | 3 - 4 | 266 | 104 | |
AK310799.1 | - | 3 - 4 | 378 | 104 | ||
AK312287.1 | - | 3 - 4 | 371 | 104 | ||
BC008426.1 | - | 3 - 4 | 376 | 4249991 | 104 | |
BX441569.2 | - | 3 - 4 | 829 | 104 | ||
CR542053.1 | - | 3 - 4 | 364 | 104 | ||
U44772.1 | - | 3 - 4 | 377 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
