Assay Details
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000310.3 | NP_000301.1 | 4 - 5 | 663 | 86 | |
XM_005271008.1 | XP_005271065.1 | 4 - 5 | 447 | 86 | ||
GenBank mRNA | AK301682.1 | - | 4 - 5 | 333 | 86 | |
AK312287.1 | - | 4 - 5 | 438 | 86 | ||
BC008426.1 | - | 4 - 5 | 443 | 4249991 | 86 | |
CR542053.1 | - | 4 - 5 | 431 | 86 | ||
U44772.1 | - | 4 - 5 | 444 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
