Assay Details
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031911.4 | NP_114117.1 | 1 - 2 | 99 | 80 | |
GenBank mRNA | AF329839.1 | - | 1 - 2 | 74 | 80 | |
DB290077.1 | - | 1 - 2 | 100 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF329839.1, NM_031911.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
