Assay Details
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002059.4 | NP_002050.1 | 4 - 5 | 595 | 120 | |
NM_022556.3 | NP_072050.1 | 4 - 5 | 550 | 120 | ||
GenBank mRNA | AB451332.1 | - | 3 - 4 | 409 | 120 | |
AB451477.1 | - | 3 - 4 | 409 | 120 | ||
AF006060.1 | - | 4 - 5 | 421 | 120 | ||
BC020760.1 | - | 4 - 5 | 516 | 4720475 | 120 | |
CR541902.1 | - | 3 - 4 | 454 | 120 | ||
CR541924.1 | - | 3 - 4 | 454 | 120 | ||
M38451.1 | - | 4 - 5 | 466 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
