Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005252.3 | NP_005243.1 | 1 - 2 | 347 | 77 | |
GenBank mRNA | AK291326.1 | - | 1 - 2 | 297 | 77 | |
AK298659.1 | - | 1 - 2 | 297 | 77 | ||
BC004490.2 | - | 1 - 2 | 268 | 3688670 | 77 | |
BX647104.1 | - | 1 - 2 | 297 | 77 | ||
CR541785.1 | - | 1 - 2 | 142 | 77 | ||
CR542267.1 | - | 1 - 2 | 142 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
