Assay Details
Gene Symbol: CLVS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283238.1 | NP_001270167.1 | 4 - 5 | 1011 | 89 | |
GenBank mRNA | AB170306.1 | - | 4 - 5 | 1051 | 89 | |
AB171660.1 | - | 4 - 5 | 1011 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
