Assay Details
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005559400.2 | XP_005559457.2 | 30 - 31 | 5964 | 68 | |
XM_015452770.1 | XP_015308256.1 | 31 - 32 | 6093 | 68 | ||
XM_015452771.1 | XP_015308257.1 | 30 - 31 | 5985 | 68 | ||
XM_015452772.1 | XP_015308258.1 | 31 - 32 | 6093 | 68 | ||
XM_015452774.1 | XP_015308260.1 | 31 - 32 | 5840 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
