Assay Details
Gene Symbol: AP5S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568431.2 | XP_005568488.1 | 1 - 2 | 525 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005568431.2 transcripts.
Set Membership: |
![]() ![]() |
