Assay Details
Gene Symbol: BNIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553176.2 | XP_005553233.1 | 9 - 10 | 2274 | 107 | |
XM_005553177.2 | XP_005553234.1 | 9 - 10 | 2272 | 107 | ||
XM_005553178.2 | XP_005553235.1 | 9 - 10 | 2278 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
