Assay Details
Gene Symbol: LIPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548941.2 | XP_005548998.1 | 5 - 6 | 753 | 117 | |
XM_005548943.2 | XP_005549000.1 | 4 - 5 | 702 | 117 | ||
XM_005548944.2 | XP_005549001.1 | 6 - 7 | 1267 | 117 | ||
XM_005548946.2 | XP_005549003.1 | 5 - 6 | 1216 | 117 | ||
XM_015446973.1 | XP_015302459.1 | 5 - 6 | 1513 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
