Assay Details
Gene Symbol: GPR171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546115.2 | XP_005546172.1 | 1 - 2 | 577 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005546115.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
