Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287677.1 | NP_001274606.1 | 1 | 376 | 110 | |
GenBank mRNA | AB171723.1 | - | 1 | 376 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
