Assay Details
Gene Symbol: CDC42EP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285016.1 | NP_001271945.1 | 2 | 749 | 72 | |
XM_005584824.2 | XP_005584881.1 | 2 | 72 | |||
XM_005584825.2 | XP_005584882.1 | 2 | 72 | |||
XM_005584827.2 | XP_005584884.1 | 2 | 72 | |||
GenBank mRNA | AB174505.1 | - | 2 | 749 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
