Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284893.1 | NP_001271822.1 | 6 - 7 | 849 | 72 | |
XM_015442119.1 | XP_015297605.1 | 7 - 8 | 796 | 72 | ||
XM_015442120.1 | XP_015297606.1 | 7 - 8 | 759 | 72 | ||
XM_015442121.1 | XP_015297607.1 | 7 - 8 | 855 | 72 | ||
XM_015442122.1 | XP_015297608.1 | 7 - 8 | 743 | 72 | ||
XM_015442123.1 | XP_015297609.1 | 7 - 8 | 773 | 72 | ||
GenBank mRNA | AB168139.1 | - | 6 - 7 | 849 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
