Assay Details
Gene Symbol: PNKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285096.1 | NP_001272025.1 | 7 - 8 | 789 | 89 | |
XM_015432086.1 | XP_015287572.1 | 7 - 8 | 836 | 89 | ||
XM_015432087.1 | XP_015287573.1 | 6 - 7 | 804 | 89 | ||
XM_015432088.1 | XP_015287574.1 | 7 - 8 | 1306 | 89 | ||
GenBank mRNA | AB172847.1 | - | 7 - 8 | 789 | 89 |
