Assay Details
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283850.1 | NP_001270779.1 | 2 - 3 | 1024 | 91 | |
GenBank mRNA | AB172745.1 | - | 2 - 3 | 1024 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
