Assay Details
Gene Symbol: MAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284558.1 | NP_001271487.1 | 1 - 2 | 179 | 67 | |
GenBank mRNA | AB171631.1 | - | 1 - 2 | 179 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
