Assay Details
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449302.1 | XP_015304788.1 | 4 - 5 | 2156 | 76 | |
XM_015449307.1 | XP_015304793.1 | 4 - 5 | 2156 | 76 | ||
XM_015449309.1 | XP_015304795.1 | 3 - 4 | 2014 | 76 | ||
XM_015449313.1 | XP_015304799.1 | 4 - 5 | 1423 | 76 | ||
XM_015449318.1 | XP_015304804.1 | 4 - 5 | 2162 | 76 | ||
XM_015449320.1 | XP_015304806.1 | 4 - 5 | 1424 | 76 | ||
XM_015449323.1 | XP_015304809.1 | 4 - 5 | 2157 | 76 |
