Assay Details
Gene Symbol: IL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557828.2 | XP_005557885.1 | 3 - 4 | 845 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
