Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319443.1 | NP_001306372.1 | 3 - 4 | 266 | 84 | |
XM_005568655.2 | XP_005568712.1 | 4 - 5 | 713 | 84 | ||
XM_015457902.1 | XP_015313388.1 | 4 - 5 | 685 | 84 | ||
GenBank mRNA | AJ320181.1 | - | 3 - 4 | 266 | 84 |
