Assay Details
Gene Symbol: ZSCAN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449702.1 | XP_015305188.1 | 2 - 3 | 599 | 103 | |
XM_015449703.1 | XP_015305189.1 | 2 - 3 | 568 | 103 | ||
XM_015449704.1 | XP_015305190.1 | 2 - 3 | 599 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
