Assay Details
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK155558.1 | - | 4 - 5 | 881 | 94 | |
AK171369.1 | - | 4 - 5 | 878 | 94 | ||
BC021653.1 | - | 4 - 5 | 901 | 94 | ||
BC127082.1 | - | 4 - 5 | 822 | 40111372 | 94 | |
DQ086834.1 | - | 4 - 5 | 861 | 94 | ||
L03290.1 | - | 3 - 4 | 708 | 94 | ||
L11600.1 | - | 3 - 4 | 708 | 94 | ||
L29006.1 | - | 3 - 4 | 1231 | 94 | ||
M62838.1 | - | 4 - 5 | 819 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
