Assay Details
Gene Symbol: Gadd45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007836.1 | NP_031862.1 | 1 - 2 | 209 | 101 | |
GenBank mRNA | AK144900.1 | - | 1 - 2 | 209 | 101 | |
BC011141.1 | - | 1 - 2 | 189 | 4035898 | 101 | |
L28177.1 | - | 1 - 2 | 209 | 101 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK144900.1, BC011141.1, L28177.1, NM_007836.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
