Assay Details
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF128221.1 | - | 3 - 4 | 579 | 74 | |
EU791538.1 | - | 7 - 8 | 1507 | 74 | ||
EU791540.1 | - | 7 - 8 | 1632 | 74 | ||
M38651.1 | - | 7 - 8 | 1571 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
