Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 12 - 13 | 1449 | 81 | |
NM_007982.2 | NP_032008.2 | 11 - 12 | 1294 | 81 | ||
GenBank mRNA | AB011499.1 | - | 9 - 10 | 982 | 81 | |
AB030035.1 | - | 9 - 10 | 982 | 81 | ||
AK079821.1 | - | 12 - 13 | 1546 | 81 | ||
AK220543.1 | - | 11 - 12 | 1294 | 81 | ||
BC030180.1 | - | 12 - 13 | 1379 | 81 | ||
JN971016.1 | - | 9 - 10 | 982 | 81 | ||
M95408.1 | - | 10 - 11 | 1091 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
