Assay Details
Gene Symbol: Fgfr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008011.2 | NP_032037.2 | 2 - 3 | 245 | 64 | |
GenBank mRNA | AK084850.1 | - | 2 - 3 | 245 | 64 | |
AK085723.1 | - | 2 - 3 | 244 | 64 | ||
AY493377.2 | - | 1 - 2 | 131 | 64 | ||
BC033313.1 | - | 2 - 3 | 432 | 5132831 | 64 | |
DQ388428.1 | - | 1 - 2 | 131 | 64 | ||
X59927.1 | - | 2 - 3 | 187 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
