Assay Details
Gene Symbol: Irak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177974.1 | NP_001171445.1 | 11 - 12 | 1709 | 63 | |
NM_001177975.1 | NP_001171446.1 | 11 - 12 | 1709 | 63 | ||
NM_001177976.1 | NP_001171447.1 | 11 - 12 | 1709 | 63 | ||
NM_008363.2 | NP_032389.2 | 11 - 12 | 1709 | 63 | ||
GenBank mRNA | AF103876.1 | - | 11 - 12 | 1709 | 63 | |
AK045958.1 | - | 11 - 12 | 1618 | 63 | ||
AK052933.1 | - | 9 - 10 | 1579 | 63 | ||
AK143694.1 | - | 11 - 12 | 1620 | 63 | ||
AK144879.1 | - | 11 - 12 | 1618 | 63 | ||
AY184362.1 | - | 11 - 12 | 1709 | 63 | ||
AY184363.1 | - | 11 - 12 | 1709 | 63 | ||
AY184364.1 | - | 11 - 12 | 1709 | 63 | ||
U56773.2 | - | 11 - 12 | 1533 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
