Assay Details
Gene Symbol: Nrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008737.2 | NP_032763.2 | 9 - 10 | 2106 | 83 | |
GenBank mRNA | AK082992.1 | - | 9 - 10 | 2159 | 83 | |
AK141964.1 | - | 9 - 10 | 1898 | 83 | ||
AK148372.1 | - | 9 - 10 | 2082 | 83 | ||
AK149752.1 | - | 4 - 5 | 687 | 83 | ||
AK159219.1 | - | 9 - 10 | 1897 | 83 | ||
BC060129.1 | - | 9 - 10 | 1886 | 6409596 | 83 | |
D50086.1 | - | 9 - 10 | 1966 | 83 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AI323974.1 | - | Chr. - - [-] on Build | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
