Assay Details
Gene Symbol: Prkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008926.4 | NP_032952.3 | 10 - 11 | 1423 | 74 | |
GenBank mRNA | AK038739.1 | - | 10 - 11 | 1407 | 74 | |
AK081422.1 | - | 10 - 11 | 1424 | 74 | ||
BC113205.1 | - | 10 - 11 | 1334 | 40047751 | 74 | |
L12460.1 | - | 10 - 11 | 1695 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
