Assay Details
Gene Symbol: Axl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190974.1 | NP_001177903.1 | 8 - 9 | 1373 | 69 | |
NM_009465.4 | NP_033491.2 | 8 - 9 | 1373 | 69 | ||
GenBank mRNA | AK033237.1 | - | 8 - 9 | 1342 | 69 | |
AK155567.1 | - | 9 - 10 | 1259 | 69 | ||
AK161294.1 | - | 1 - 2 | 46 | 69 | ||
AK170010.1 | - | 8 - 9 | 1347 | 69 | ||
BC008551.1 | - | 3 - 4 | 463 | 69 | ||
BC046618.1 | - | 8 - 9 | 1210 | 6494383 | 69 | |
BC050914.1 | - | 8 - 9 | 1181 | 6418252 | 69 | |
BC058230.1 | - | 9 - 10 | 1261 | 6313662 | 69 | |
X63535.1 | - | 8 - 9 | 1122 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
