Assay Details
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009603.1 | NP_033733.1 | 11 - 12 | 1345 | 70 | |
GenBank mRNA | AK006411.1 | - | 2 - 3 | 237 | 70 | |
X55718.1 | - | 11 - 12 | 1345 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901659 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901660 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901658 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180470 | X55718.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180470 | NM_009603.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720812 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197947 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874731 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158812 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180470 | X55718.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180470 | NM_009603.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482820 | X55718.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482820 | NM_009603.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P522601 | X55718.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P522601 | NM_009603.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P522601 | AF032863.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901699 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901689 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901697 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901693 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901691 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901690 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901712 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901685 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901674 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901680 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901702 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901708 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901705 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901698 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901701 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901679 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901716 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901675 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901695 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901673 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901706 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901672 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901677 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901714 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901704 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901687 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901682 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901710 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158812 | - | - |
Whole Mouse Genome G4122A | A_52_P522601 | X55718.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P522601 | NM_009603.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P522601 | AF032863.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P482820 | X55718.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P482820 | NM_009603.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130982 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158812 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09255651 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09255641 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09255647 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09255650 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09666157 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09666156 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901660 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901659 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09255650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09614328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09666156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901658 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09183481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09255647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09686305 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09643148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27901675 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10387916 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387911 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387912 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387914 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387918 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387910 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387913 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387921 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387920 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387917 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387915 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387919 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420560_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10387909 | - | - |
Mouse Genome 430 2.0 Array | 1420560_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420560_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11448_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11448_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100028640.mm.1 | TC1100003104.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028640.mm.1 | NM_009603.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028640.mm.1 | NM_009603.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028639.mm.1 | TC1100003104.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028639.mm.1 | NM_009603.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028639.mm.1 | NM_009603.1 | Refseq |
Mouse Expression Set 430 | 1420560_at | - | - |
Mouse Expression Set 430 | 1420560_PM_at | - | - |
Murine Genome U74v2 Set | 92965_at | - | - |
Murine Genome U74v2 Set | 92967_r_at | - | - |
Murine Genome U74v2 Set | 92965_PM_at | - | - |
Murine Genome U74v2 Set | 92966_PM_g_at | - | - |
Murine Genome U74v2 Set | 92966_g_at | - | - |
Murine Genome U74v2 Set | 92967_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1420560_at | - | - |
Mouse Genome 430A 2.0 Array | 1420560_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420560_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420560_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 355219 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3190170 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3190170 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003190170 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706323 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2706323 | - | - |
mouse-6 v1.1 (Illumina) | scl39993.11.1_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3190170 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39993.11.1_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190170 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2706326 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2706326 | - | - |
