Assay Details
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271566.1 | NP_001258495.1 | 7 - 8 | 1148 | 106 | |
NM_009863.3 | NP_033993.2 | 7 - 8 | 979 | 106 | ||
GenBank mRNA | AB018575.1 | - | 7 - 8 | 980 | 106 | |
AB018576.1 | - | 6 - 7 | 790 | 106 | ||
AB018577.1 | - | 6 - 7 | 790 | 106 | ||
AJ007661.1 | - | 7 - 8 | 931 | 106 | ||
AK145921.1 | - | 7 - 8 | 905 | 106 | ||
AK146009.1 | - | 7 - 8 | 1150 | 106 | ||
BC062149.1 | - | 7 - 8 | 909 | 106 | ||
BC072666.1 | - | 7 - 8 | 920 | 30611704 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
