Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
TC0X00000941.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177937.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177937.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177938.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177938.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177939.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177939.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177940.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177940.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177941.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177943.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177943.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177944.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_001177944.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_010099.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006105.mm.1 |
NM_010099.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
TC0X00000941.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177937.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177937.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177938.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177938.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177939.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177939.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177940.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177940.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177941.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177943.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177943.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177944.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_001177944.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_010099.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006106.mm.1 |
NM_010099.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
TC0X00000941.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177937.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177937.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177938.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177938.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177939.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177939.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177940.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177940.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177941.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177943.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177943.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177944.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_001177944.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_010099.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006100.mm.1 |
NM_010099.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
TC0X00000941.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177937.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177937.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177938.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177938.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177939.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177939.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177940.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177940.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177941.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177942.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177943.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177943.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177944.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_001177944.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_010099.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00006108.mm.1 |
NM_010099.2 |
Refseq |