Assay Details
Gene Symbol: Lgr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010195.2 | NP_034325.2 | 6 - 7 | 1001 | 108 | |
GenBank mRNA | AF110818.1 | - | 6 - 7 | 714 | 108 | |
AK047873.1 | - | 6 - 7 | 983 | 108 | ||
AK132387.1 | - | 6 - 7 | 1001 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
