Assay Details
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122899.1 | NP_001116371.1 | 16 - 17 | 3039 | 81 | |
NM_010704.2 | NP_034834.1 | 16 - 17 | 3039 | 81 | ||
NM_146146.2 | NP_666258.2 | 16 - 17 | 3039 | 81 | ||
GenBank mRNA | AK136231.1 | - | 2 - 3 | 772 | 81 | |
AK140785.1 | - | 16 - 17 | 2541 | 81 | ||
AK143989.1 | - | 16 - 17 | 2728 | 81 | ||
AK155891.1 | - | 16 - 17 | 2580 | 81 | ||
BC082551.1 | - | 16 - 17 | 2296 | 30621706 | 81 | |
U42467.1 | - | 16 - 17 | 2547 | 81 | ||
U46135.1 | - | 16 - 17 | 2547 | 81 | ||
U49106.1 | - | 15 - 16 | 2487 | 81 | ||
U49107.1 | - | 3 - 4 | 500 | 81 | ||
U49108.1 | - | 3 - 4 | 500 | 81 | ||
U49109.1 | - | 15 - 16 | 2487 | 81 | ||
U52915.1 | - | 16 - 17 | 3001 | 81 | ||
U58861.1 | - | 16 - 17 | 2569 | 81 | ||
U58862.1 | - | 16 - 17 | 2579 | 81 | ||
U58863.1 | - | 16 - 17 | 2569 | 81 | ||
Y10296.1 | - | 15 - 16 | 2495 | 81 | ||
Y10298.1 | - | 15 - 16 | 2495 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
