Assay Details
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141813.1 | - | 1 - 2 | 236 | 87 | |
AK164484.1 | - | 1 - 2 | 237 | 87 | ||
BC063768.1 | - | 1 - 2 | 218 | 87 | ||
CJ066818.1 | - | 1 - 2 | 253 | 87 | ||
U14166.1 | - | 1 - 2 | 259 | 87 | ||
U15159.1 | - | 1 - 2 | 234 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U14166.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
