Assay Details
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139231.1 | - | 2 - 3 | 2674 | 80 | |
AK141813.1 | - | 9 - 10 | 1329 | 80 | ||
AK170166.1 | - | 8 - 9 | 1104 | 80 | ||
AK172083.1 | - | 8 - 9 | 1088 | 80 | ||
BC063768.1 | - | 9 - 10 | 1309 | 80 | ||
U14166.1 | - | 9 - 10 | 1352 | 80 | ||
U15159.1 | - | 9 - 10 | 1324 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
