Assay Details
Gene Symbol: Nos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010927.3 | NP_035057.1 | 14 - 15 | 1871 | 75 | |
GenBank mRNA | AF065919.1 | - | 14 - 15 | 1868 | 75 | |
AF065920.1 | - | 14 - 15 | 1868 | 75 | ||
AF065921.2 | - | 14 - 15 | 1868 | 75 | ||
AF065922.2 | - | 13 - 14 | 1741 | 75 | ||
AF065923.2 | - | 13 - 14 | 1741 | 75 | ||
AY090567.1 | - | 13 - 14 | 1691 | 75 | ||
BC062378.1 | - | 14 - 15 | 1880 | 4978648 | 75 | |
M84373.1 | - | 14 - 15 | 1872 | 75 | ||
M87039.1 | - | 14 - 15 | 1943 | 75 | ||
M92649.1 | - | 14 - 15 | 1879 | 75 | ||
U43428.1 | - | 14 - 15 | 1759 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
