Assay Details
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011103.3 | NP_035233.1 | 4 - 5 | 614 | 67 | |
GenBank mRNA | AB201453.1 | - | 1 - 2 | 216 | 67 | |
AB201454.1 | - | 1 - 2 | 216 | 67 | ||
AB201455.1 | - | 1 - 2 | 216 | 67 | ||
AB201456.1 | - | 1 - 2 | 216 | 67 | ||
AF251036.1 | - | 3 - 4 | 377 | 67 | ||
AK144231.1 | - | 4 - 5 | 619 | 67 | ||
AK168768.1 | - | 4 - 5 | 609 | 67 | ||
AY545076.1 | - | 3 - 4 | 377 | 67 | ||
BC051416.1 | - | 4 - 5 | 578 | 6515302 | 67 | |
M69042.1 | - | 4 - 5 | 609 | 67 | ||
X60304.1 | - | 3 - 4 | 390 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB201453.1, AB201454.1, AB201455.1, AB201456.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
