Assay Details
Gene Symbol: Sema6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272024.1 | NP_001258953.1 | 14 - 15 | 1746 | 63 | |
NM_011351.2 | NP_035481.1 | 11 - 12 | 1314 | 63 | ||
NR_073549.1 | 12 - 13 | 1639 | 63 | |||
NR_073550.1 | 12 - 13 | 1653 | 63 | |||
NR_073551.1 | 13 - 14 | 1702 | 63 | |||
GenBank mRNA | AB013729.1 | - | 11 - 12 | 1279 | 63 | |
AF363972.1 | - | 10 - 11 | 1227 | 63 | ||
AF363973.1 | - | 11 - 12 | 1347 | 63 | ||
AK031131.1 | - | 12 - 13 | 1551 | 63 | ||
AK129459.1 | - | 6 - 7 | 790 | 63 | ||
AK161042.1 | - | 6 - 7 | 793 | 63 | ||
AK165085.1 | - | 12 - 13 | 1566 | 63 | ||
BC051979.1 | - | 13 - 14 | 1608 | 63 | ||
BC059007.1 | - | 14 - 15 | 1657 | 63 | ||
BC083186.1 | - | 7 - 8 | 2730 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
