Assay Details
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK133647.1 | - | 2319 | 64 | ||
M37890.1 | - | 5 - 6 | 2258 | 64 | ||
S56585.1 | - | 5 - 6 | 2337 | 64 | ||
X53779.1 | - | 5 - 6 | 2290 | 64 | ||
X59411.1 | - | 5 - 6 | 2289 | 64 | ||
X59592.1 | - | 5 - 6 | 2378 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
