Assay Details
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY605711.1 | - | 7 - 8 | 931 | 87 | |
BC144799.1 | - | 7 - 8 | 987 | 9053347 | 87 | |
BC145743.1 | - | 7 - 8 | 987 | 40131046 | 87 | |
M74425.1 | - | 7 - 8 | 985 | 84 | ||
X72395.1 | - | 7 - 8 | 949 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
