Assay Details
Gene Symbol: Prpf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013830.2 | NP_038858.2 | 3 - 4 | 1572 | 97 | |
GenBank mRNA | AB516304.1 | - | 3 - 4 | 1424 | 97 | |
AF033663.1 | - | 2 - 3 | 862 | 97 | ||
AF283466.1 | - | 3 - 4 | 1570 | 97 | ||
AK078611.1 | - | 3 - 4 | 1538 | 97 | ||
AK083926.1 | - | 3 - 4 | 1456 | 97 | ||
BC042521.1 | - | 4 - 5 | 855 | 97 | ||
BC080864.1 | - | 3 - 4 | 1544 | 97 | ||
BC141272.1 | - | 3 - 4 | 1498 | 9055884 | 97 | |
BC141273.1 | - | 3 - 4 | 1498 | 9055885 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
