Assay Details
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013865.2 | NP_038893.1 | 8 - 9 | 689 | 82 | |
NM_180956.1 | NP_851287.1 | 8 - 9 | 689 | 82 | ||
GenBank mRNA | AB033922.1 | - | 8 - 9 | 603 | 82 | |
AK002272.1 | - | 7 - 8 | 603 | 82 | ||
AK004689.1 | - | 8 - 9 | 635 | 82 | ||
AK036277.1 | - | 8 - 9 | 614 | 82 | ||
AK037487.1 | - | 8 - 9 | 615 | 82 | ||
AK154927.1 | - | 8 - 9 | 656 | 82 | ||
AK218281.1 | - | 4 - 5 | 259 | 82 | ||
BC018504.1 | - | 8 - 9 | 623 | 4500970 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
