Assay Details
Gene Symbol: Ikbke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019777.3 | NP_062751.2 | 4 - 5 | 573 | 76 | |
GenBank mRNA | AB016589.1 | - | 2 - 3 | 261 | 76 | |
AK088580.1 | - | 4 - 5 | 575 | 76 | ||
AK149911.1 | - | 4 - 5 | 562 | 76 | ||
AK153046.1 | - | 4 - 5 | 574 | 76 | ||
CT010206.1 | - | 2 - 3 | 227 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
