Assay Details
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023580.4 | NP_076069.2 | 6 - 7 | 1402 | 75 | |
GenBank mRNA | AF131197.1 | - | 6 - 7 | 1344 | 75 | |
AK028478.1 | - | 6 - 7 | 1403 | 75 | ||
BC071215.1 | - | 6 - 7 | 1366 | 4196138 | 75 | |
U18084.1 | - | 2 - 3 | 505 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
